Learn More
Cayman Chemical O-2545 (hydrochloride), Purity: ≥98%, CAS: 874745-43-4, 10 mg
SDP
Catalog No. 502888246
Encompass_Preferred
A potent water-soluble agonist of CB1 and CB2 receptors with Ki values of 1.5 and 0.32 nM, respectively; when dissolved in saline, is highly efficacious in mouse behavioral models when administered either intravenously or intracerebroventricularly
Catalog No. 50-288-8246 Supplier Cayman Chemical Supplier No. 1000919510 MG
A potent water-soluble agonist of CB1 and CB2 receptors with Ki values of 1.5 and 0.32 nM, respectively; when dissolved in saline, is highly efficacious in mouse behavioral models when administered either intravenously or intracerebroventricularly