Learn More
Medchemexpress LLC HY-18772 5mg Medchemexpress, O4I2 CAS:165682-93-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY187725MG
Encompass_Preferred
Medchemexpress, HY-18772 5mg O4I2 CAS:165682-93-9 O4I2 is a potent Oct3/4 inducer. O4I2 induces the expression of pluripotent-associated genes Lin28, Sox2 and Nanog, and suppresses Rex1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2088