Learn More
Waters Corp Bottle Domino, 55 mm, 10x Bottle Capacity, 1x2 Format, for Andrew+ Robot
Catalog No. NC3243227 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 55 mm 10x Bottle Domino is a machine-readable consumable holder specifically designed to accommodate up to 10 large-volume bottles. It functions as an interface between various consumable types and the Andrew+ pipetting robot, facilitating seamless automation of laboratory workflows. Equipped with QR codes for labware information and ARUCO codes for spatial positioning, Dominos are essential components in any Andrew+ automation setup.
  • Holds up to 10 large-volume bottles, enabling automated laboratory workflows with the Andrew+ pipetting robot.
  • 1x2 format, occupying two horizontal positions on the robot workbench.
  • Designed for use with the Andrew+ pipetting robot.
  • Features QR codes for critical labware information and ARUCO codes for spatial positioning, making it machine-readable.
Catalog No. NC3243227 Supplier Waters Corp Supplier No. 186010318
The 55 mm 10x Bottle Domino is a machine-readable consumable holder specifically designed to accommodate up to 10 large-volume bottles. It functions as an interface between various consumable types and the Andrew+ pipetting robot, facilitating seamless automation of laboratory workflows. Equipped with QR codes for labware information and ARUCO codes for spatial positioning, Dominos are essential components in any Andrew+ automation setup.
  • Holds up to 10 large-volume bottles, enabling automated laboratory workflows with the Andrew+ pipetting robot.
  • 1x2 format, occupying two horizontal positions on the robot workbench.
  • Designed for use with the Andrew+ pipetting robot.
  • Features QR codes for critical labware information and ARUCO codes for spatial positioning, making it machine-readable.