Learn More
Enzo Life Sciences O6-Benzylguanine (10mg). CAS: 19916-73-5
SDP
Supplier:  Enzo Life Sciences ALX480019M010
Encompass_Preferred
Effective substrate and efficient inactivator of DNA alkyltransferase. Purity: ≥98% (TLC). Appearance: White to off-white powder. Solubility: Soluble in methanol (20mg/ml). Long Term Storage: Ambient.
Catalog No. 50-200-9305