Learn More
Selleck Chemical LLC O6-Benzylguanine 25mg 19916-73-5 O6-BG
SDP
Supplier:  Selleck Chemical LLC S365825MG
Encompass_Preferred
O6-Benzylguanine (O6-BG) is a potent O6-alkylguanine DNA alkyltransferase (AGT) inactivator. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8327