Learn More
Cayman Chemical O6-Benzylguanine, Purity: ≥98%, CAS: 19916-73-5, 10 mg
SDP
Catalog No. 502970503
Encompass_Preferred
A guanine analog with antineoplastic activity; irreversibly inactivates AGT, with 40% reduction in alkyltransferase activity achieved at 0.06 µM with recombinant human AGT; effective in vivo as well as in vitro
Catalog No. 50-297-0503 Supplier Cayman Chemical Supplier No. 1633210 MG
A guanine analog with antineoplastic activity; irreversibly inactivates AGT, with 40% reduction in alkyltransferase activity achieved at 0.06 µM with recombinant human AGT; effective in vivo as well as in vitro