Learn More
Cayman Chemical O6-Benzylguanine, Purity: ≥98%, CAS: 19916-73-5, 50 mg
SDP
Catalog No. 502970476
Encompass_Preferred
A guanine analog with antineoplastic activity; irreversibly inactivates AGT, with 40% reduction in alkyltransferase activity achieved at 0.06 µM with recombinant human AGT; effective in vivo as well as in vitro
Catalog No. 50-297-0476 Supplier Cayman Chemical Supplier No. 1633250 MG
A guanine analog with antineoplastic activity; irreversibly inactivates AGT, with 40% reduction in alkyltransferase activity achieved at 0.06 µM with recombinant human AGT; effective in vivo as well as in vitro