Learn More
Cayman Chemical O6-methylguanine-DNA Methyltr
SDP
Catalog No. 502951084
Encompass_Preferred
Recombinant N-terminal His-tagged protein expressed in E. coli • Amino acids: 2-238 (full length) • MW: 26.8 kDa
Catalog No. 50-295-1084 Supplier Cayman Chemical Supplier No. 11176100 UG
Recombinant N-terminal His-tagged protein expressed in E. coli • Amino acids: 2-238 (full length) • MW: 26.8 kDa