Learn More
STEMCELL Technologies OAC1, Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 72602
Encompass
OAC1 is an OCT-4-activating compound that activates expression through the OCT-4 gene promoter (Li et al.). Oct-4 (POU5F1) is a transcription factor that is critically involved in the self-renewal of pluripotent stem cells, and its expression is commonly used as a marker for pluripotency.REPROGRAMMING· Enhances the reprogramming efficiency of mouse embryonic fibroblasts transfected with OCT4, SOX2, KLF4, and c-MYC (Li et al.).MAINTENANCE AND SELF-RENEWAL· Mediates ex vivo expansion of cord blood CD34+ hematopoietic stem and progenitor cells (Huang et al.).
Catalog No. 50-197-5948