Learn More
Medchemexpress LLC HY-12884 10mg Medchemexpress, OAC2 CAS:6019-39-2 Purity:>98%
SDP
Catalog No. 502081188
Encompass_Preferred
Medchemexpress, HY-12884 10mg OAC2 CAS:6019-39-2 OAC2 is an Oct4-activating compound which activates expression through the Oct4 gene promoter. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1188 Supplier Medchemexpress LLC Supplier No. HY1288410MG
Medchemexpress, HY-12884 10mg OAC2 CAS:6019-39-2 OAC2 is an Oct4-activating compound which activates expression through the Oct4 gene promoter. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.