Learn More
TARGETMOL CHEMICALS INC OAC2 25MG
SDP
Catalog No. 502577245
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OAC2 is an Oct4 activator which activates expression through the Oct4 gene promoter; enhances reprogramming efficiency by increasing the rate of production of induced pluripotent stem cells (iPSCs) from embryonic fibroblasts; an analog of OAC1. purity: 99%
Catalog No. 50-257-7245 Supplier TARGETMOL CHEMICALS INC Supplier No. T201925MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OAC2 is an Oct4 activator which activates expression through the Oct4 gene promoter; enhances reprogramming efficiency by increasing the rate of production of induced pluripotent stem cells (iPSCs) from embryonic fibroblasts; an analog of OAC1. purity: 99%