Learn More
Electron Microscopy Sciences Oak Ridge Style Centrifuge Tubes, 12/PK
SDP
Catalog No. 502090808
Encompass_Preferred
Oakridge Style re-usable Centrifuge Tubes are economic choice for high speed refrigerated or non-refrigerated centrifugation. When filled to 80% of their nominal capacity the tubes withstand up to 50,000g . They are molded from durable, chemical resistant polypropylene and have round bottoms and consistent wall thicknesses. Oakridge style tubes are autoclavable and re-usable and are designed with leakproof linerless closures. The have excellent chemical resistance and an attached screw cap. Capacity: 50ml.
Catalog No. 50-209-0808 Supplier Electron Microscopy Sciences Supplier No. 6304503
Oakridge Style re-usable Centrifuge Tubes are economic choice for high speed refrigerated or non-refrigerated centrifugation. When filled to 80% of their nominal capacity the tubes withstand up to 50,000g . They are molded from durable, chemical resistant polypropylene and have round bottoms and consistent wall thicknesses. Oakridge style tubes are autoclavable and re-usable and are designed with leakproof linerless closures. The have excellent chemical resistance and an attached screw cap. Capacity: 50ml.