Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Oak Table with Plain Apron and Solid Phenolic Top: Electrical Option
SCI_ED

Catalog No. S41915 Shop All Diversified Spaces Products
Change view
Click to view available options
Dimensions (L x W):
21 x 28 in.
21 x 54 in.
21 x 60 in.
21 x 72 in.
24 x 48 in.
24 x 54 in.
24 x 60 in.
24 x 72 in.
30 x 48 in.
30 x 54 in.
30 x 60 in. (76.2 x 152.4 cm)
30 x 72 in.
36 x 48 in.
36 x 54 in.
36 x 60 in.
36 x 72 in.
Height (English):
30 in.
34 in.
36 in.
36 product options available for selection
Product selection table with 36 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Height (English)
S41915 24 x 72 in. 34 in.
S42107 24 x 60 in. 30 in.
S42114 24 x 60 in. 34 in.
S42711 30 x 48 in. 36 in.
S42121 24 x 60 in. 36 in.
S47456 24 x 48 in. 30 in.
S47466 24 x 48 in. 36 in.
S47461 24 x 48 in. 34 in.
S47607 30 x 48 in. 30 in.
S47680 30 x 54 in. 30 in.
S47960 30 x 72 in. 30 in.
S47694 30 x 54 in. 36 in.
S47821 30 x 60 in. (76.2 x 152.4 cm) 36 in.
S47967 30 x 72 in. 34 in.
S48076 21 x 28 in. 36 in.
S48069 21 x 28 in. 34 in.
S48154 36 x 48 in. 34 in.
S48161 36 x 48 in. 36 in.
S48233 21 x 54 in. 34 in.
S48318 36 x 54 in. 34 in.
S48226 21 x 54 in. 30 in.
S48311 36 x 54 in. 30 in.
S48240 21 x 54 in. 36 in.
S48325 36 x 54 in. 36 in.
S42784 21 x 28 in. 30 in.
S48436 24 x 54 in. 34 in.
S48556 21 x 60 in. 30 in.
S48443 24 x 54 in. 36 in.
S48563 21 x 60 in. 34 in.
S48570 21 x 60 in. 36 in.
S41812 36 x 72 in. 30 in.
S48641 36 x 60 in. 30 in.
S48648 36 x 60 in. 34 in.
S48709 21 x 72 in. 30 in.
S41819 36 x 72 in. 34 in.
S48716 21 x 72 in. 34 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 36 options available.
36 options
Catalog No. S41915 Supplier Diversified Spaces™ Supplier No. P7304K34E
Add to Cart
Add to Cart

Ideal for high school and above with low temperature use. Diversified Spaces™ Oak Tables with Phelonic Tops are designed for use as counter tops, pegboards and reagent racks.

  • 1 in. Phenolic Table Surface
  • 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Includes UL rated electrical that can be hard wired or portable cord connected by certified electrician
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required; legs need to be attached
  • Solid Oak Construction

Specifications

Product Type Plain Apron Table
Color Oak
Dimensions (L x W) 24 x 72 in.
Style Plain Apron
Certifications/Compliance SEFA compliant and MAS Certified Green™
Adjustable No
For Use With (Application) High school and above and with low temperature use
Includes Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only
Finish Chemical resistant, earth-friendly UV
Height (English) 34 in.
Height (Metric) 86.36 cm
Load Bearing Capacity (English) 500 lb.
Load Bearing Capacity (Metric) 226.79 kg
Top Style Solid Phenolic
Material Red Oak
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.