Learn More
Cole-Parmer Buffer Solution, pH 10.01; 1000 mL
Supplier:  Cole-Parmer 0594262
Encompass_Preferred
All Oakton pH buffers are traceable to NIST standard reference materials. Buffer solution lets you calibrate pH instruments to ensure accuracy. If unopened, shelf life is 2 years. USA buffer value of pH 10.01.
Catalog No. NC1769482