Learn More
Cole-Parmer Oakton Buffer Pack; 500 mL of each pH 4.01, 7.00, and 10.01
Catalog No. NC1762202 Shop All Cole-Parmer Products
Encompass_Preferred
• Oakton pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations • Manufactured under ISO 9000 quality guidelines and traceable to NIST standards • If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days • Store buffers away from direct sunlight.
Catalog No. NC1762202 Supplier Cole-Parmer Supplier No. 0594210
• Oakton pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations • Manufactured under ISO 9000 quality guidelines and traceable to NIST standards • If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days • Store buffers away from direct sunlight.