Learn More
Calibration Kit for TDSTestr™ and ECTestr™ Waterproof Microprocessor TDS/Conductivity Meters, Oakton™
Catalog No. 13200241
Change view
Click to view available options
Quantity:
20 mL/Ea
Catalog No. Quantity
13-200-241 20 mL/Ea
1 options
This item is not returnable. View return policy
Catalog No. 13-200-241 Supplier Oakton Supplier No. 3566170
This item is not returnable. View return policy
s. Includes everything needed for convenient calibration, measurement and storage of waterproof testers.
TDSTestr/ECTestrs Calibration Kit. For use with both Standard and Plus Series Models. Contains two each of TDS Singles calibration solution pouches (20mL each of 447, 1413, 2764, and 15,000μS solutions), rinse water, sample jar, instructions, screwdriver, and protective carrying case that organizes meter and material. Accuracy of ±1% at 25°C. Testers not included.
Compliance
Calibration solutions made with NIST traceable materials
Specifications
Description TDSTestr Calibration Kit
For Use With (Equipment) TDSTestr
Quantity 20 mL/Ea