Learn More
Cole-Parmer Oakton pH Solutions Pack; pH 4.01/7.00/10.00, Storage/Cleaning, 500 mL each
Catalog No. NC9741067 Shop All Cole-Parmer Products
Encompass_Preferred
Oakton pH Solutions Pack; pH 4.01/7.00/10.00, Storage/Cleaning, 500 mL each, Oakton pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrationsCleaning solution removes protein, oil, and grease contaminantsperfect for general-cleaning use too!Storage solution provides proper hydration to ensure accuracy and stability
Catalog No. NC9741067 Supplier Cole-Parmer Supplier No. 0594215
Oakton pH Solutions Pack; pH 4.01/7.00/10.00, Storage/Cleaning, 500 mL each, Oakton pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrationsCleaning solution removes protein, oil, and grease contaminantsperfect for general-cleaning use too!Storage solution provides proper hydration to ensure accuracy and stability