Learn More
Grainger Oakton All-in-One pH Electrode, Epoxy, ATC, 1 ea
Catalog No. 502785939
Encompass_Preferred
This all-in-one combination pH electrode features a durable epoxy body with a built-in temperature sensor for convenient, simultaneous pH and temperature measurements. Designed for reliability and ease of use, it utilizes a double-junction design, which helps prevent clogging and contamination in demanding environments such as those containing heavy metals or organics. The probe includes an integrated 76.2 cm (30 in.) cable with a BNC connector for seamless integration with compatible pH meters.
  • Double-junction reference design for enhanced resistance to fouling
  • Epoxy body construction for rugged, long-lasting performance
  • Built-in automatic temperature compensation (ATC) sensor
  • Suitable for diverse applications including industrial, environmental, and laboratory testing
  • Dimensions: 14.6 cm (5.75 in.) length and 12 mm (0.5 in.) diameter
Catalog No. 50-278-5939 Supplier Grainger Supplier No. 192NE8
This all-in-one combination pH electrode features a durable epoxy body with a built-in temperature sensor for convenient, simultaneous pH and temperature measurements. Designed for reliability and ease of use, it utilizes a double-junction design, which helps prevent clogging and contamination in demanding environments such as those containing heavy metals or organics. The probe includes an integrated 76.2 cm (30 in.) cable with a BNC connector for seamless integration with compatible pH meters.
  • Double-junction reference design for enhanced resistance to fouling
  • Epoxy body construction for rugged, long-lasting performance
  • Built-in automatic temperature compensation (ATC) sensor
  • Suitable for diverse applications including industrial, environmental, and laboratory testing
  • Dimensions: 14.6 cm (5.75 in.) length and 12 mm (0.5 in.) diameter