Learn More
Sigma Aldrich Fine Chemicals Biosciences OaktonR AcornTM series pH1EA
Catalog No. 502908726
Encompass
Reads pH mV and temperatureThree-point pH calibration w/auto buffer recognitionAutomatic Temperature Compensation (ATC)Toggle between pH and temperature in CPush-button operation hold function auto-off3-year warranty on meterMeter includes pH electrode w/BNC connector temperature probe operating instructions and batteries.pH calibration up to 3 points 4.01 7.00 10.01 increments.Temperature calibrationOffset 0.1C incrementsmV calibration /-20 mV
Catalog No. 50-290-8726 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5278071EA
Reads pH mV and temperatureThree-point pH calibration w/auto buffer recognitionAutomatic Temperature Compensation (ATC)Toggle between pH and temperature in CPush-button operation hold function auto-off3-year warranty on meterMeter includes pH electrode w/BNC connector temperature probe operating instructions and batteries.pH calibration up to 3 points 4.01 7.00 10.01 increments.Temperature calibrationOffset 0.1C incrementsmV calibration /-20 mV