Learn More
eMolecules​ 106-40-1 | 4-Bromoaniline | Oakwood Chemical | MFCD00007822 | 172.025 | C6H6BrN | 99.000 | Nc1ccc(Br)cc1 | 1kg | 537663435
Catalog No. 5000130549 Shop All eMolecules Inc. Products
Encompass_Preferred
4-Bromoaniline | Oakwood Chemical | 106-40-1 | MFCD00007822 | 172.025 | C6H6BrN | 99.000 | Nc1ccc(Br)cc1 | 1kg | 537663435
Catalog No. 50-001-30549 Supplier eMolecules​ Supplier No. 537663435
4-Bromoaniline | Oakwood Chemical | 106-40-1 | MFCD00007822 | 172.025 | C6H6BrN | 99.000 | Nc1ccc(Br)cc1 | 1kg | 537663435