Learn More
Medchemexpress LLC OARV-771 | 2683008-37-7 | 98.8% | C49H59ClN8O8S2 | 10MG
SDP
Catalog No. 5000261765
Encompass_Preferred
OARV-771 is a VHL-based BET degrader (PROTAC) that induces proteasomal degradation of BET family proteins BRD2, BRD3, and BRD4. It demonstrates low-nanomolar degradation potency in cellular assays and is supplied as a solid with high chemical purity for research use.
  • Low-nanomolar degradation potency (DC50: Brd2 ~1 nM, Brd3 ~4 nM, Brd4 ~6 nM).
  • VHL-based PROTAC mechanism for targeted protein degradation.
  • Improved cell permeability for cellular studies.
  • High chemical purity suitable for biochemical and cell-based assays.
  • Solid form for convenient handling and formulation.
  • Recommended sealed cold storage to maintain stability.
Catalog No. 50-002-61765 Supplier Medchemexpress LLC Supplier No. HY14526410MG
OARV-771 is a VHL-based BET degrader (PROTAC) that induces proteasomal degradation of BET family proteins BRD2, BRD3, and BRD4. It demonstrates low-nanomolar degradation potency in cellular assays and is supplied as a solid with high chemical purity for research use.
  • Low-nanomolar degradation potency (DC50: Brd2 ~1 nM, Brd3 ~4 nM, Brd4 ~6 nM).
  • VHL-based PROTAC mechanism for targeted protein degradation.
  • Improved cell permeability for cellular studies.
  • High chemical purity suitable for biochemical and cell-based assays.
  • Solid form for convenient handling and formulation.
  • Recommended sealed cold storage to maintain stability.