Learn More
Medchemexpress LLC OARV-771 | 2683008-37-7 | 98.8% | C49H59ClN8O8S2 | 25 MG
SDP
Catalog No. 5000354595
Encompass_Preferred
OARV-771 is a VHL-based BET degrader (PROTAC) with improved cell permeability. It shows DC50s of 6 nM for Brd4, 1 nM for Brd2, and 4 nM for Brd3. It also exhibits an antiproliferative effect with an EC50 value of 4 nM in MV4;11 cells.
  • VHL-based BET degrader (PROTAC)
  • Enhanced cell permeability
  • Demonstrates DC50s of 6 nM for Brd4, 1 nM for Brd2, and 4 nM for Brd3
  • Exhibits an antiproliferative effect with an EC50 value of 4 nM in MV4;11 cells
Catalog No. 50-003-54595 Supplier Medchemexpress LLC Supplier No. 25MGHY145264
OARV-771 is a VHL-based BET degrader (PROTAC) with improved cell permeability. It shows DC50s of 6 nM for Brd4, 1 nM for Brd2, and 4 nM for Brd3. It also exhibits an antiproliferative effect with an EC50 value of 4 nM in MV4;11 cells.
  • VHL-based BET degrader (PROTAC)
  • Enhanced cell permeability
  • Demonstrates DC50s of 6 nM for Brd4, 1 nM for Brd2, and 4 nM for Brd3
  • Exhibits an antiproliferative effect with an EC50 value of 4 nM in MV4;11 cells