Learn More
Aviva Systems Biology Corp OAS1 Antibody - N-terminal region (ARP40401_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40401_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OAS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OAS1. Alternate names: IFI-4, OIAS, OIASIApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57434