Learn More
Aviva Systems Biology Corp Oas1g Antibody - N-terminal region (ARP40402_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40402_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Oas1g. It was validated on Western Blot. . Alternate names: AI449562, L2, Mmu-L, Mmu-L2, Oas1a, Oias-1, Oias1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-869