Learn More
Aviva Systems Biology Corp OAS2 Antibody - middle region (ARP40852_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40852_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OAS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OAS2. Alternate names: MGC78578ApplicationWB. Species reactivity: Cow, Dog, Human, Pig, Rabbit
Catalog No. 50-014-57639