Learn More
MSC Ecolab Floor Cleaner, For Cleaning Floors, Clear/Light Yellow, 2.5 gal
Catalog No. 502816358
Encompass
This heavy-duty, self-foaming, chlorinated alkaline cleaner is designed for use in kitchens to effectively remove soap film, grease, oil, and other common soils. It is not intended for use as a sanitizer or disinfectant.
  • Heavy-duty, self-foaming, chlorinated alkaline cleaner
  • Designed for use in kitchens
  • Effectively removes soap film, grease, oil, and other common soils
  • Not intended for use as a sanitizer or disinfectant
Catalog No. 50-281-6358 Supplier MSC Supplier No. 01229053
This heavy-duty, self-foaming, chlorinated alkaline cleaner is designed for use in kitchens to effectively remove soap film, grease, oil, and other common soils. It is not intended for use as a sanitizer or disinfectant.
  • Heavy-duty, self-foaming, chlorinated alkaline cleaner
  • Designed for use in kitchens
  • Effectively removes soap film, grease, oil, and other common soils
  • Not intended for use as a sanitizer or disinfectant