Learn More
Waters Corp Waters Vac Cartridge, GCB/WAX, PFAS Analysis, 60 μm
Catalog No. NC3314962 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Vac Cartridges combine a mixed-mode, Weak Anion-eXchange (WAX), reversed-phase, water-wettable polymer with graphitized carbon black (GCB). Designed for high purity and consistency, they enable low-level quantification of per- and polyfluoroalkyl substances (PFAS) in environmental samples across various matrices.
  • Delivers high purity and consistency for diverse sample analysis.
  • Facilitates low-level quantification of per- and polyfluoroalkyl substances (PFAS).
  • Provides GCB clean-up, as described in EPA Draft Method 1633.
  • Integrates WAX and GCB for time savings and a simplified workflow.
  • Ensures PFAS extraction and concentration without cross-contamination.
Catalog No. NC3314962 Supplier Waters Corp Supplier No. 186011113
These Vac Cartridges combine a mixed-mode, Weak Anion-eXchange (WAX), reversed-phase, water-wettable polymer with graphitized carbon black (GCB). Designed for high purity and consistency, they enable low-level quantification of per- and polyfluoroalkyl substances (PFAS) in environmental samples across various matrices.
  • Delivers high purity and consistency for diverse sample analysis.
  • Facilitates low-level quantification of per- and polyfluoroalkyl substances (PFAS).
  • Provides GCB clean-up, as described in EPA Draft Method 1633.
  • Integrates WAX and GCB for time savings and a simplified workflow.
  • Ensures PFAS extraction and concentration without cross-contamination.