Learn More
Waters Corp Waters SPE Cartridge, GCB/WAX, PFAS Analysis, 6 cc
Catalog No. NC3314957 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis WAX for PFAS Analysis products feature a mixed-mode, weak anion-eXchange, reversed-phase, water-wettable polymer. Designed for high purity and consistency in sample analysis across various matrices including water, soil, and food. They enable low-level quantification of per- and polyfluoroalkyl substances (PFAS) in environmental samples, addressing regulatory concerns.
  • Mixed-mode, weak anion-eXchange, reversed-phase, water-wettable polymer.
  • Low-level PFAS quantification in environmental samples.
  • GCB clean-up for complex matrices, adhering to EPA Draft Method 1633.
  • Single cartridge combines WAX and GCB clean-up, simplifying workflow.
  • QC pre-screening for 32 PFAS prevents cross-contamination.
  • Application-specific QC testing ensures accurate results.
  • Ideal for EPA Draft Method 1633 for solids, biosolids, and tissue.
Catalog No. NC3314957 Supplier Waters Corp Supplier No. 186011112
Oasis WAX for PFAS Analysis products feature a mixed-mode, weak anion-eXchange, reversed-phase, water-wettable polymer. Designed for high purity and consistency in sample analysis across various matrices including water, soil, and food. They enable low-level quantification of per- and polyfluoroalkyl substances (PFAS) in environmental samples, addressing regulatory concerns.
  • Mixed-mode, weak anion-eXchange, reversed-phase, water-wettable polymer.
  • Low-level PFAS quantification in environmental samples.
  • GCB clean-up for complex matrices, adhering to EPA Draft Method 1633.
  • Single cartridge combines WAX and GCB clean-up, simplifying workflow.
  • QC pre-screening for 32 PFAS prevents cross-contamination.
  • Application-specific QC testing ensures accurate results.
  • Ideal for EPA Draft Method 1633 for solids, biosolids, and tissue.