Learn More
Waters Corp Waters Vac Cartridge, HLB, 3 cc, 540 mg, 60 μm
Catalog No. 50179322 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 3 cc Oasis HLB cartridges utilize a universal polymeric reversed-phase sorbent for extracting a wide range of acidic, basic, and neutral compounds. The water-wettable sorbent maintains high retention and excellent recoveries even when dry, simplifying sample preparation. With a 60 μm particle size, these syringe-barrel cartridges are ideal for viscous samples, compatible with vacuum manifolds and automated SPE instruments.
  • Offers broad pH stability (0 to 14) and solvent compatibility.
  • Provides exceptional retention for polar molecules.
  • Hydrophobic retention capacity is three times higher than typical silica-based SPE sorbents.
  • Water-wettable sorbent maintains performance even if the bed runs dry.
  • Enhances recovery and repeatability of SPE procedures due to higher retention capacity.
  • Constructed with ultra-clean polypropylene syringe barrel and frits.
Catalog No. 50-179-322 Supplier Waters Corp Supplier No. 186004134
The 3 cc Oasis HLB cartridges utilize a universal polymeric reversed-phase sorbent for extracting a wide range of acidic, basic, and neutral compounds. The water-wettable sorbent maintains high retention and excellent recoveries even when dry, simplifying sample preparation. With a 60 μm particle size, these syringe-barrel cartridges are ideal for viscous samples, compatible with vacuum manifolds and automated SPE instruments.
  • Offers broad pH stability (0 to 14) and solvent compatibility.
  • Provides exceptional retention for polar molecules.
  • Hydrophobic retention capacity is three times higher than typical silica-based SPE sorbents.
  • Water-wettable sorbent maintains performance even if the bed runs dry.
  • Enhances recovery and repeatability of SPE procedures due to higher retention capacity.
  • Constructed with ultra-clean polypropylene syringe barrel and frits.