Learn More
Waters Corp Waters Corporation μElution Plate, Oasis HLB, 2 mg Sorbent, 30 μm
Catalog No. 50818606 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis HLB μElution plate is a 96-well solid phase extraction plate designed for analyte enrichment and cleanup of sample volumes from 10 μL to 375 μL. Each well contains 2 mg of Oasis HLB sorbent, a universal polymeric reversed-phase sorbent. It allows elution in just 25 μL, providing sensitive and reproducible results without evaporation and reconstitution, and enables up to 15-fold sample enrichment.
  • Enables extraction of a wide range of acidic, basic, and neutral compounds from various matrices using a simple, generic protocol.
  • Compatible with most liquid-handling robotic systems for automated, high throughput SPE.
  • Offers superior retention and recovery for both polar and non-polar compounds.
  • Water-wettable sorbents eliminate the need for repeat preparation due to sorbent bed drying.
Catalog No. 50-818-606 Supplier Waters Corp Supplier No. 186001828BA
Add to Cart
Add to Cart
The Oasis HLB μElution plate is a 96-well solid phase extraction plate designed for analyte enrichment and cleanup of sample volumes from 10 μL to 375 μL. Each well contains 2 mg of Oasis HLB sorbent, a universal polymeric reversed-phase sorbent. It allows elution in just 25 μL, providing sensitive and reproducible results without evaporation and reconstitution, and enables up to 15-fold sample enrichment.
  • Enables extraction of a wide range of acidic, basic, and neutral compounds from various matrices using a simple, generic protocol.
  • Compatible with most liquid-handling robotic systems for automated, high throughput SPE.
  • Offers superior retention and recovery for both polar and non-polar compounds.
  • Water-wettable sorbents eliminate the need for repeat preparation due to sorbent bed drying.