Learn More
Waters Corp Bulk Media, Oasis HLB, 250 g, 30 μm, Reversed Phase
Catalog No. 502391602 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This bulk media is an Oasis HLB SPE sorbent, with a 30 μm particle size. It is a universal polymeric reversed-phase sorbent, designed for the extraction of a wide range of acidic, basic, and neutral compounds from various matrices using a simple, generic protocol.
  • Universal polymeric reversed-phase sorbent for extracting a wide range of acidic, basic, and neutral compounds.
  • Utilizes a simple, generic protocol for extraction.
  • Water wettability ensures higher retention and excellent recoveries even if the sorbent runs dry.
  • Eliminates the need for extraordinary steps to prevent sorbent beds from drying out during critical steps prior to sample loading.
  • Copolymer particle substrate.
  • Pore size of 80 Å.
  • No silanol activity.
  • pH range of 0 to 14 pH.
Catalog No. 50-239-1602 Supplier Waters Corp Supplier No. 186007550
This bulk media is an Oasis HLB SPE sorbent, with a 30 μm particle size. It is a universal polymeric reversed-phase sorbent, designed for the extraction of a wide range of acidic, basic, and neutral compounds from various matrices using a simple, generic protocol.
  • Universal polymeric reversed-phase sorbent for extracting a wide range of acidic, basic, and neutral compounds.
  • Utilizes a simple, generic protocol for extraction.
  • Water wettability ensures higher retention and excellent recoveries even if the sorbent runs dry.
  • Eliminates the need for extraordinary steps to prevent sorbent beds from drying out during critical steps prior to sample loading.
  • Copolymer particle substrate.
  • Pore size of 80 Å.
  • No silanol activity.
  • pH range of 0 to 14 pH.