Learn More
Waters Corp Waters Vac Cartridge, HLB, 12 cc, 500 mg, 60 μm
Catalog No. 50780076 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Vac Cartridge contains Oasis HLB, a universal polymeric reversed-phase sorbent designed for solid-phase extraction (SPE) of a wide range of acidic, basic, and neutral compounds from various matrices. Featuring a hydrophilic-lipophilic balanced (HLB) chemistry and a 60 μm particle size, this water-wettable cartridge offers exceptional performance and simplified methods, making it a gold standard in SPE.
  • Water wettable, maintaining retention and recoveries even if the sorbent runs dry.
  • Stable across a wide pH range, from 0 to 14.
  • Eliminates equilibration and conditioning steps, reducing solvent consumption and sample preparation time.
  • Provides high sensitivity and reduced matrix effects for LC-MS/MS analyses.
  • Offers unsurpassed selectivity and high SPE recovery for cleaner samples.
Catalog No. 50-780-076 Supplier Waters Corp Supplier No. 186000116
This Vac Cartridge contains Oasis HLB, a universal polymeric reversed-phase sorbent designed for solid-phase extraction (SPE) of a wide range of acidic, basic, and neutral compounds from various matrices. Featuring a hydrophilic-lipophilic balanced (HLB) chemistry and a 60 μm particle size, this water-wettable cartridge offers exceptional performance and simplified methods, making it a gold standard in SPE.
  • Water wettable, maintaining retention and recoveries even if the sorbent runs dry.
  • Stable across a wide pH range, from 0 to 14.
  • Eliminates equilibration and conditioning steps, reducing solvent consumption and sample preparation time.
  • Provides high sensitivity and reduced matrix effects for LC-MS/MS analyses.
  • Offers unsurpassed selectivity and high SPE recovery for cleaner samples.