Learn More
Waters Corp Oasis Vac Cartridge, 1 cc, 30 mg Sorbent, 30 μm
Catalog No. 50785843 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 1 cc cartridges contain the HLB sorbent, a universal polymeric reversed-phase sorbent developed for extracting a wide range of acidic, basic, and neutral compounds from various matrices using a simple, generic protocol. This sorbent is water wettable, maintaining higher retention and excellent recoveries even if the sorbent runs dry. The cartridge is designed for automated Solid Phase Extraction (SPE) on the Gilson ASPEC.
  • Universal polymeric reversed-phase sorbent for extracting a wide range of acidic, basic, and neutral compounds.
  • Water wettable, maintaining higher retention and excellent recoveries even if the sorbent runs dry.
  • Designed for automated Solid Phase Extraction (SPE) on the Gilson ASPEC.
Catalog No. 50-785-843 Supplier Waters Corp Supplier No. WAT058882
These 1 cc cartridges contain the HLB sorbent, a universal polymeric reversed-phase sorbent developed for extracting a wide range of acidic, basic, and neutral compounds from various matrices using a simple, generic protocol. This sorbent is water wettable, maintaining higher retention and excellent recoveries even if the sorbent runs dry. The cartridge is designed for automated Solid Phase Extraction (SPE) on the Gilson ASPEC.
  • Universal polymeric reversed-phase sorbent for extracting a wide range of acidic, basic, and neutral compounds.
  • Water wettable, maintaining higher retention and excellent recoveries even if the sorbent runs dry.
  • Designed for automated Solid Phase Extraction (SPE) on the Gilson ASPEC.