Learn More
Waters Corp Oasis Vac Cartridge, HLB, 1 cc, 30 mg, 30 μm
Catalog No. 50785991 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 1 cc cartridges contain a universal polymeric reversed-phase sorbent, water wettable and designed for extracting a wide range of compounds. They maintain high retention and excellent recoveries even if the sorbent runs dry and are suitable for vacuum manifolds and automated SPE instruments.
  • Universal polymeric reversed-phase sorbent.
  • Water-wettable for high retention and recovery.
  • Compatible with vacuum manifolds and automated SPE.
  • Superior capacity and enhanced retention for polar analytes.
  • Stable across pH 0 to 14 and in various solvents.
  • 3x higher hydrophobic retention than silica-based sorbents.
  • Simplifies SPE by eliminating conditioning/equilibration steps.
  • Reduces matrix effects for high sensitivity LC-MS/MS.
  • Ensures high SPE recovery, quality, and reproducibility.
  • Reduces solvent consumption and sample preparation time.
Catalog No. 50-785-991 Supplier Waters Corp Supplier No. WAT094225
The 1 cc cartridges contain a universal polymeric reversed-phase sorbent, water wettable and designed for extracting a wide range of compounds. They maintain high retention and excellent recoveries even if the sorbent runs dry and are suitable for vacuum manifolds and automated SPE instruments.
  • Universal polymeric reversed-phase sorbent.
  • Water-wettable for high retention and recovery.
  • Compatible with vacuum manifolds and automated SPE.
  • Superior capacity and enhanced retention for polar analytes.
  • Stable across pH 0 to 14 and in various solvents.
  • 3x higher hydrophobic retention than silica-based sorbents.
  • Simplifies SPE by eliminating conditioning/equilibration steps.
  • Reduces matrix effects for high sensitivity LC-MS/MS.
  • Ensures high SPE recovery, quality, and reproducibility.
  • Reduces solvent consumption and sample preparation time.