Learn More
Waters Corp Waters Vac Cartridge, HLB, 30 μm, 1 cc
Catalog No. 50179321 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • High retention capacity for diverse analytes.
  • Maintains retention even if sorbent bed runs dry.
  • Enhances SPE durability; reduces sample preparation repeats.
  • Universal sorbent for acidic, neutral, and basic compounds.
  • Offers pH stability from 0 to 14 pH.
  • Excellent polar molecule retention.
  • Hydrophobic retention capacity 3x higher than silica-based SPE sorbents.
  • Designed for vacuum manifolds and automated SPE instruments.
  • Enables extraction of diverse chemicals using generic techniques.
  • Increases analyte retention and minimizes breakthrough.
  • Improves recovery and overall SPE repeatability.
This item is not returnable. View return policy
Catalog No. 50-179-321 Supplier Waters Corp Supplier No. 186003908
  • High retention capacity for diverse analytes.
  • Maintains retention even if sorbent bed runs dry.
  • Enhances SPE durability; reduces sample preparation repeats.
  • Universal sorbent for acidic, neutral, and basic compounds.
  • Offers pH stability from 0 to 14 pH.
  • Excellent polar molecule retention.
  • Hydrophobic retention capacity 3x higher than silica-based SPE sorbents.
  • Designed for vacuum manifolds and automated SPE instruments.
  • Enables extraction of diverse chemicals using generic techniques.
  • Increases analyte retention and minimizes breakthrough.
  • Improves recovery and overall SPE repeatability.