Learn More
Waters Corp Oasis Vac Cartridge, 3 cc, 400 mg Sorbent, 60 μm
Catalog No. 50201491 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis HLB Vac Cartridge is a multi-purpose, reversed-phase sorbent with a water-wettable copolymer stable from pH 0–14. It provides analyte specificity and increased sensitivity for Solid Phase Extraction (SPE), allowing analytes to be retained even when loaded directly in an aqueous sample solution, offering excellent recoveries and higher retention.
  • Water-wettable copolymer stable from pH 0–14.
  • Eliminates condition and equilibration steps, reducing overall sample preparation time.
  • Reduces solvent consumption by up to 70% and sample preparation time by 40%.
  • Offers high recoveries and retention for diverse compounds.
  • Hydrophilic-lipophilic balance retains both polar and reversed-phase compounds.
  • Higher retention capacity improves recovery and reproducibility of SPE methods.
Catalog No. 50-201-491 Supplier Waters Corp Supplier No. 186003849
The Oasis HLB Vac Cartridge is a multi-purpose, reversed-phase sorbent with a water-wettable copolymer stable from pH 0–14. It provides analyte specificity and increased sensitivity for Solid Phase Extraction (SPE), allowing analytes to be retained even when loaded directly in an aqueous sample solution, offering excellent recoveries and higher retention.
  • Water-wettable copolymer stable from pH 0–14.
  • Eliminates condition and equilibration steps, reducing overall sample preparation time.
  • Reduces solvent consumption by up to 70% and sample preparation time by 40%.
  • Offers high recoveries and retention for diverse compounds.
  • Hydrophilic-lipophilic balance retains both polar and reversed-phase compounds.
  • Higher retention capacity improves recovery and reproducibility of SPE methods.