Learn More
Waters Corp Waters Oasis HLB Vac Cartridge, 6 cc, 150 mg Sorbent, 30 μm, 30/pk
Catalog No. 50781717 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 6 cc Oasis HLB cartridges contain a universal polymeric reversed-phase sorbent for extracting various compounds. Its water-wettable nature ensures high retention and excellent recoveries.
  • Made using a specific ratio of two monomers: hydrophilic N-vinylpyrrolidone and lipophilic divinylbenzene
  • Hydrophilic-Lipophilic-Balanced, water-wettable reversed-phase universal sorbent
  • Effective for acidic, neutral, and basic compounds
  • Stability at pH extremes and in a wide range of solvents
  • Extraordinary retention of polar compounds
  • Relative hydrophobic retention capacity three times higher than traditional silica-based SPE sorbents
  • Maintains higher retention and excellent recoveries even if the sorbent runs dry
  • Designed for use with vacuum manifolds and automated SPE instruments
Catalog No. 50-781-717 Supplier Waters Corp Supplier No. 186003365
These 6 cc Oasis HLB cartridges contain a universal polymeric reversed-phase sorbent for extracting various compounds. Its water-wettable nature ensures high retention and excellent recoveries.
  • Made using a specific ratio of two monomers: hydrophilic N-vinylpyrrolidone and lipophilic divinylbenzene
  • Hydrophilic-Lipophilic-Balanced, water-wettable reversed-phase universal sorbent
  • Effective for acidic, neutral, and basic compounds
  • Stability at pH extremes and in a wide range of solvents
  • Extraordinary retention of polar compounds
  • Relative hydrophobic retention capacity three times higher than traditional silica-based SPE sorbents
  • Maintains higher retention and excellent recoveries even if the sorbent runs dry
  • Designed for use with vacuum manifolds and automated SPE instruments