Learn More
Waters Corp Waters Corporation Vac Cartridge, Oasis HLB, 6 cc, 150 mg, 60 μm
Catalog No. 50781728 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These cartridges feature Oasis HLB sorbent, a universal polymeric reversed-phase sorbent for extracting diverse compounds from various matrices. Its water-wettable nature ensures consistent high retention and excellent recoveries, even if the sorbent bed dries out. The 60 μm particle size is ideal for viscous samples, and the syringe-barrel design allows compatibility with vacuum manifolds and automated SPE instruments.
  • Universal polymeric reversed-phase for diverse compound extraction.
  • Water wettable for consistent retention, even when dry.
  • 60 μm particle size, ideal for viscous samples.
  • Syringe-barrel format for vacuum manifolds and automated SPE.
  • Stable across extreme pH (0 to 14) and wide solvent range.
  • High retention for polar and hydrophobic compounds.
  • Manufactured to ISO 9001 & cGMP standards for reproducibility.
Catalog No. 50-781-728 Supplier Waters Corp Supplier No. 186003379
These cartridges feature Oasis HLB sorbent, a universal polymeric reversed-phase sorbent for extracting diverse compounds from various matrices. Its water-wettable nature ensures consistent high retention and excellent recoveries, even if the sorbent bed dries out. The 60 μm particle size is ideal for viscous samples, and the syringe-barrel design allows compatibility with vacuum manifolds and automated SPE instruments.
  • Universal polymeric reversed-phase for diverse compound extraction.
  • Water wettable for consistent retention, even when dry.
  • 60 μm particle size, ideal for viscous samples.
  • Syringe-barrel format for vacuum manifolds and automated SPE.
  • Stable across extreme pH (0 to 14) and wide solvent range.
  • High retention for polar and hydrophobic compounds.
  • Manufactured to ISO 9001 & cGMP standards for reproducibility.