Learn More
Waters Corp Waters Oasis HLB Vac Cartridge, 6 cc, 500 mg Sorbent, 60 μm
Catalog No. 50780075 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 6 cc Oasis HLB cartridges contain a universal polymeric reversed-phase sorbent for extracting a wide range of compounds. The water wettable sorbent ensures high retention and excellent recoveries even if it runs dry. A 60 μm particle size is recommended for viscous samples, and the cartridges are designed for vacuum manifolds and automated SPE instruments.
  • High capacity, low elution volumes.
  • Excellent recovery for polar metabolites.
  • Detection limits of 100 ng/L or better.
  • Faster flow times, less vacuum needed.
  • Extraordinarily clean barrels and frits.
  • Proprietary cleaning process for minimal interferences.
  • Stable at pH extremes, retains polar compounds, hydrophobic.
Catalog No. 50-780-075 Supplier Waters Corp Supplier No. 186000115
These 6 cc Oasis HLB cartridges contain a universal polymeric reversed-phase sorbent for extracting a wide range of compounds. The water wettable sorbent ensures high retention and excellent recoveries even if it runs dry. A 60 μm particle size is recommended for viscous samples, and the cartridges are designed for vacuum manifolds and automated SPE instruments.
  • High capacity, low elution volumes.
  • Excellent recovery for polar metabolites.
  • Detection limits of 100 ng/L or better.
  • Faster flow times, less vacuum needed.
  • Extraordinarily clean barrels and frits.
  • Proprietary cleaning process for minimal interferences.
  • Stable at pH extremes, retains polar compounds, hydrophobic.