Learn More
Waters Corp Waters Oasis HLB Vac Cartridge, 10 mg, 30 μm, ASPC
Catalog No. 50179317 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Oasis HLB Vac Cartridge with Gilson ASPC adapter is designed for solid phase extraction applications. It features HLB chemistry and is water wettable, ensuring reliable performance across a broad pH range.
  • Designed for solid phase extraction
  • Reversed phase mode for various applications
  • Water wettable for consistent performance
  • Mass spec compatibility ensures analytical integrity
  • Wide pH range from 0 to 14
  • HLB chemistry for broad analyte retention
  • Vac cartridge format for ease of use
  • 30 μm particle size
  • 10 mg sorbent weight
  • Copolymer sorbent substrate
  • Constructed from polypropylene material
  • 1 cc barrel size
Catalog No. 50-179-317 Supplier Waters Corp Supplier No. 186000988
The Waters Oasis HLB Vac Cartridge with Gilson ASPC adapter is designed for solid phase extraction applications. It features HLB chemistry and is water wettable, ensuring reliable performance across a broad pH range.
  • Designed for solid phase extraction
  • Reversed phase mode for various applications
  • Water wettable for consistent performance
  • Mass spec compatibility ensures analytical integrity
  • Wide pH range from 0 to 14
  • HLB chemistry for broad analyte retention
  • Vac cartridge format for ease of use
  • 30 μm particle size
  • 10 mg sorbent weight
  • Copolymer sorbent substrate
  • Constructed from polypropylene material
  • 1 cc barrel size