Learn More
Waters Corp WATERS Oasis HLB Direct Connect Column, 80Å, 2.1 mm × 15 mm
Catalog No. 50780762 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed for rugged, reproducible, and ultra-fast on-line solid phase extraction (SPE), utilizing a universal polymeric sorbent optimized for extracting a wide range of analytes from various matrices. It is intended for use with appropriate narrow-bore analytical columns.
  • Designed for rugged and reproducible on-line SPE.
  • Provides ultra-fast on-line SPE.
  • Contains a universal polymeric sorbent.
  • Optimized for the extraction of a wide range of analytes.
  • Extracts analytes from various matrices.
  • Features a direct connect hardware configuration for convenient column connection.
Catalog No. 50-780-762 Supplier Waters Corp Supplier No. 186001792
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This column is designed for rugged, reproducible, and ultra-fast on-line solid phase extraction (SPE), utilizing a universal polymeric sorbent optimized for extracting a wide range of analytes from various matrices. It is intended for use with appropriate narrow-bore analytical columns.
  • Designed for rugged and reproducible on-line SPE.
  • Provides ultra-fast on-line SPE.
  • Contains a universal polymeric sorbent.
  • Optimized for the extraction of a wide range of analytes.
  • Extracts analytes from various matrices.
  • Features a direct connect hardware configuration for convenient column connection.