Learn More
Waters Corp Waters Oasis HLB Vac Cartridge, 20 cc, 1 g, 60 μm
Catalog No. 50818659 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 20 cc Oasis HLB cartridges, featuring a 60 μm particle size, utilize a universal polymeric reversed-phase sorbent for extracting a wide range of compounds. The sorbent is water wettable, ensuring high retention and excellent recoveries even if it dries out. These syringe-barrel-type cartridges are ideal for vacuum manifolds and automated SPE instruments.
  • Made from hydrophilic N-vinylpyrrolidone and lipophilic divinylbenzene monomers.
  • Stable at pH extremes and in various solvents.
  • Offers extraordinary retention of polar compounds.
  • Three times higher hydrophobic retention than traditional silica-based SPE.
  • Improves recovery and reproducibility by retaining more analytes.
  • Water wettable for consistent performance.
  • Suitable for viscous samples.
Catalog No. 50-818-659 Supplier Waters Corp Supplier No. 186000117
The 20 cc Oasis HLB cartridges, featuring a 60 μm particle size, utilize a universal polymeric reversed-phase sorbent for extracting a wide range of compounds. The sorbent is water wettable, ensuring high retention and excellent recoveries even if it dries out. These syringe-barrel-type cartridges are ideal for vacuum manifolds and automated SPE instruments.
  • Made from hydrophilic N-vinylpyrrolidone and lipophilic divinylbenzene monomers.
  • Stable at pH extremes and in various solvents.
  • Offers extraordinary retention of polar compounds.
  • Three times higher hydrophobic retention than traditional silica-based SPE.
  • Improves recovery and reproducibility by retaining more analytes.
  • Water wettable for consistent performance.
  • Suitable for viscous samples.