Learn More
Waters Corp Oasis HLB 96-well Plate, 5 mg Sorbent per Well, 30 µm, 1/pk
Catalog No. 50819039 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis HLB 96-well plate contains the Oasis HLB sorbent, which is a universal polymeric reversed-phase sorbent that was developed for the extraction of a wide range of acidic, basic, and neutral compounds from various matrices using a simple, generic protocol. The unique balance of hydrophobicity and water-wettability of the Oasis HLB sorbent means you will never have to worry about poor results if individual wells of the 96-well plate dry out during the critical steps prior to sample loading. The Oasis HLB 96-well plate is designed to be used on many manifold configurations and most robotic liquid handling systems.
Catalog No. 50-819-039 Supplier Waters Corp Supplier No. 186000309
The Oasis HLB 96-well plate contains the Oasis HLB sorbent, which is a universal polymeric reversed-phase sorbent that was developed for the extraction of a wide range of acidic, basic, and neutral compounds from various matrices using a simple, generic protocol. The unique balance of hydrophobicity and water-wettability of the Oasis HLB sorbent means you will never have to worry about poor results if individual wells of the 96-well plate dry out during the critical steps prior to sample loading. The Oasis HLB 96-well plate is designed to be used on many manifold configurations and most robotic liquid handling systems.