Learn More
Waters Corp Waters Flangeless Vac Cartridge, Oasis HLB, 1 cc, 30 μm
Catalog No. 50780783 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 1 cc flangeless cartridges contain Oasis HLB, a water wettable, universal polymeric reversed-phase sorbent. They are designed for extracting a wide range of compounds using a simple, generic protocol, and their flangeless configuration allows close packing in automated SPE systems.
  • Sorbent: Oasis HLB, universal polymeric reversed-phase.
  • Water wettable, ensuring consistent retention and recoveries even if dry.
  • Flangeless design for close packing in automated SPE systems.
  • Sorbent weight: 30 mg.
  • Particle size: 30 μm.
  • pH range: 0 to 14.
  • Material: Polypropylene.
  • Barrel size: 1 cc.
  • Mass spec compatible.
Catalog No. 50-780-783 Supplier Waters Corp Supplier No. 186001879
These 1 cc flangeless cartridges contain Oasis HLB, a water wettable, universal polymeric reversed-phase sorbent. They are designed for extracting a wide range of compounds using a simple, generic protocol, and their flangeless configuration allows close packing in automated SPE systems.
  • Sorbent: Oasis HLB, universal polymeric reversed-phase.
  • Water wettable, ensuring consistent retention and recoveries even if dry.
  • Flangeless design for close packing in automated SPE systems.
  • Sorbent weight: 30 mg.
  • Particle size: 30 μm.
  • pH range: 0 to 14.
  • Material: Polypropylene.
  • Barrel size: 1 cc.
  • Mass spec compatible.