Learn More
Waters Corp Oasis Flangeless Vac Cartridge, 1 cc, 10 mg Sorbent, 30 μm
Catalog No. 50201604 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 1 cc flangeless cartridges contain a universal polymeric reversed-phase sorbent, developed for the extraction of a wide range of acidic, basic, and neutral compounds from various matrices using a simple, generic protocol.
  • Water wettable sorbent maintains high retention and excellent recoveries even if it runs dry.
  • Eliminates the need for special measures to prevent sorbent beds from drying out before sample loading.
  • Flangeless configuration allows cartridges to be closely packed for processing in specialized automated SPE systems.
  • HLB chemistry.
  • No endcapping.
  • Mass spectrometry compatible.
  • Reversed phase mode.
  • Copolymer sorbent substrate.
  • pH range of 0 to 14.
  • Constructed from polypropylene material.
Catalog No. 50-201-604 Supplier Waters Corp Supplier No. 186006339
The 1 cc flangeless cartridges contain a universal polymeric reversed-phase sorbent, developed for the extraction of a wide range of acidic, basic, and neutral compounds from various matrices using a simple, generic protocol.
  • Water wettable sorbent maintains high retention and excellent recoveries even if it runs dry.
  • Eliminates the need for special measures to prevent sorbent beds from drying out before sample loading.
  • Flangeless configuration allows cartridges to be closely packed for processing in specialized automated SPE systems.
  • HLB chemistry.
  • No endcapping.
  • Mass spectrometry compatible.
  • Reversed phase mode.
  • Copolymer sorbent substrate.
  • pH range of 0 to 14.
  • Constructed from polypropylene material.