Learn More
Waters Corp Waters Flangeless Vac Cartridge, HLB, 3 cc, 540 mg, 60 μm
Catalog No. 50782044 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 3 cc flangeless cartridges contain the Oasis HLB sorbent, a universal polymeric reversed-phase sorbent for extracting a wide range of acidic, basic, and neutral compounds. This sorbent is water wettable, maintaining retention and excellent recoveries even if it runs dry, simplifying sample preparation by eliminating the need to prevent sorbent beds from drying out. The 60 μm particle size is recommended for viscous samples, and the flangeless configuration allows for close packing in automated SPE systems.
  • Universal polymeric reversed-phase sorbent for a wide range of compounds
  • Simple, generic extraction protocol
  • Water wettable sorbent maintains retention & excellent recoveries even if dry
  • 60 μm particle size for viscous samples
  • Flangeless configuration for automated SPE systems
Catalog No. 50-782-044 Supplier Waters Corp Supplier No. 186003852
The 3 cc flangeless cartridges contain the Oasis HLB sorbent, a universal polymeric reversed-phase sorbent for extracting a wide range of acidic, basic, and neutral compounds. This sorbent is water wettable, maintaining retention and excellent recoveries even if it runs dry, simplifying sample preparation by eliminating the need to prevent sorbent beds from drying out. The 60 μm particle size is recommended for viscous samples, and the flangeless configuration allows for close packing in automated SPE systems.
  • Universal polymeric reversed-phase sorbent for a wide range of compounds
  • Simple, generic extraction protocol
  • Water wettable sorbent maintains retention & excellent recoveries even if dry
  • 60 μm particle size for viscous samples
  • Flangeless configuration for automated SPE systems