Learn More
Waters Corp WATERS CORPORATION Oasis HLB Flangeless Cartridge, 3 cc, 60 mg, 30 μm
Catalog No. 50780784 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 3 cc flangeless cartridges contain HLB sorbent, a universal polymeric reversed-phase sorbent for extracting various acidic, basic, and neutral compounds. This water wettable sorbent maintains high retention and excellent recoveries even if it runs dry. The flangeless configuration allows for close packing in automated SPE systems.
  • Universal polymeric reversed-phase sorbent.
  • Water wettable for high retention and recovery.
  • Flangeless design for automated SPE systems.
  • Mixed-mode functionality for analyte specificity.
  • Eliminates equilibration and conditioning steps.
  • Stable from pH 0 to 14.
  • Exceptional product quality and reproducibility.
Catalog No. 50-780-784 Supplier Waters Corp Supplier No. 186001880
The 3 cc flangeless cartridges contain HLB sorbent, a universal polymeric reversed-phase sorbent for extracting various acidic, basic, and neutral compounds. This water wettable sorbent maintains high retention and excellent recoveries even if it runs dry. The flangeless configuration allows for close packing in automated SPE systems.
  • Universal polymeric reversed-phase sorbent.
  • Water wettable for high retention and recovery.
  • Flangeless design for automated SPE systems.
  • Mixed-mode functionality for analyte specificity.
  • Eliminates equilibration and conditioning steps.
  • Stable from pH 0 to 14.
  • Exceptional product quality and reproducibility.