Learn More
Waters Corp Waters Oasis HLB Glass Vac Cartridge, 5 cc, 200 mg, 60 μm
Catalog No. 50780455 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These glass cartridges are optimized for trace analysis, detecting substances at parts-per-trillion (PPT) levels. Each lot is tested for bisphenol A and other phenols and phthalates, ensuring accurate endocrine disruptor analysis. They feature an ultra-clean glass syringe with Teflon frit and utilize a Hydrophilic-Lipophilic-Balanced, water-wettable reversed-phase universal sorbent, made from N-vinylpyrrolidone and divinylbenzene, suitable for acidic, neutral, and basic compounds.
  • Stability at pH extremes and in a wide range of solvents.
  • Extraordinary retention of polar compounds.
  • Hydrophobic retention capacity three times higher than traditional silica-based sorbents.
  • Water-wettable sorbent maintains retention even if the bed runs dry.
  • Higher retention capacity leads to improved recovery and reproducibility.
  • Provides cleaner samples with less time and effort.
Catalog No. 50-780-455 Supplier Waters Corp Supplier No. 186000683
These glass cartridges are optimized for trace analysis, detecting substances at parts-per-trillion (PPT) levels. Each lot is tested for bisphenol A and other phenols and phthalates, ensuring accurate endocrine disruptor analysis. They feature an ultra-clean glass syringe with Teflon frit and utilize a Hydrophilic-Lipophilic-Balanced, water-wettable reversed-phase universal sorbent, made from N-vinylpyrrolidone and divinylbenzene, suitable for acidic, neutral, and basic compounds.
  • Stability at pH extremes and in a wide range of solvents.
  • Extraordinary retention of polar compounds.
  • Hydrophobic retention capacity three times higher than traditional silica-based sorbents.
  • Water-wettable sorbent maintains retention even if the bed runs dry.
  • Higher retention capacity leads to improved recovery and reproducibility.
  • Provides cleaner samples with less time and effort.