Learn More
Waters Corp Oasis HLB Online Cartridge Column, 80Å, 5 µm, 3.9 mm X 20 mm, 1/pk
Supplier:  Waters Corp 186001413
Encompass_Preferred
The Oasis HLB on-line cartridge column is designed for rugged, reproducible, and ultra-fast on-line SPE. This cartridge column contains the universal polymeric Oasis HLB sorbent, which is optimized for the extraction of a wide range of analytes from various matrices. This on-line cartridge column is designed to be used with the appropriate Waters narrow-bore analytical column, such as XSelect, XBridge, SunFire, Atlantis, XTerra, or Symmetry column. The Oasis HLB on-line cartridge column fits into a Sentry holder that features a finger-tight fitting for fast, convenient replacement.
Catalog No. 50-780-690