Learn More
Waters Corp Waters Cartridge, HLB, 80Å, 30 μm, 10 mm
Catalog No. 50466265 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis HLB on-line cartridges for Symbiosis/Prospekt-2 systems are designed for rugged, reproducible, and ultra-fast on-line SPE in the Spark Holland Symbiosis and Prospekt-2 systems. Each cartridge contains 2.5 mg of Oasis HLB, which is a universal polymeric reversed-phase sorbent for the extraction of a wide range of acidic, basic, and neutral compounds from various matrices.
  • Designed for rugged, reproducible, and ultra-fast on-line SPE in Spark Holland Symbiosis and Prospekt-2 systems.
  • Each cartridge contains 2.5 mg of Oasis HLB, a universal polymeric reversed-phase sorbent.
  • Suitable for extraction of a wide range of acidic, basic, and neutral compounds from various matrices.
  • An 800-series tray contains 96 cartridges.
  • Can be directly loaded into Automatic Cartridge Exchange (ACE) or ACE Feeder units without a stainless steel tray holder/adaptor.
Catalog No. 50-466-265 Supplier Waters Corp Supplier No. 186005781
The Oasis HLB on-line cartridges for Symbiosis/Prospekt-2 systems are designed for rugged, reproducible, and ultra-fast on-line SPE in the Spark Holland Symbiosis and Prospekt-2 systems. Each cartridge contains 2.5 mg of Oasis HLB, which is a universal polymeric reversed-phase sorbent for the extraction of a wide range of acidic, basic, and neutral compounds from various matrices.
  • Designed for rugged, reproducible, and ultra-fast on-line SPE in Spark Holland Symbiosis and Prospekt-2 systems.
  • Each cartridge contains 2.5 mg of Oasis HLB, a universal polymeric reversed-phase sorbent.
  • Suitable for extraction of a wide range of acidic, basic, and neutral compounds from various matrices.
  • An 800-series tray contains 96 cartridges.
  • Can be directly loaded into Automatic Cartridge Exchange (ACE) or ACE Feeder units without a stainless steel tray holder/adaptor.